3,5-Bis(methylthio)benzotrifluoride ≥95%

Supplier: AOBChem USA

26011-500MG 26011-1G 26011-5G 26011-25G
77324-740EA 176.13 USD
77324-740 77346-510 77361-066 77384-384
3,5-Bis(methylthio)benzotrifluoride ≥95%
3,5-Bis(methylthio)benzotrifluoride

Precaución: for Research Use Only

Formula: C₉H₉F₃S₂
MDL Number: MFCD24848706
CAS Number: 1449008-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR