6-Chloro-2-fluoro-3-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

26004-500MG 26004-1G 26004-5G 26004-25G
77324-736EA 116.39 USD
77324-736 77343-482 77361-062 77372-832
6-Chloro-2-fluoro-3-iodobenzaldehyde ≥97%
6-Chloro-2-fluoro-3-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃ClFIO
MW: 284.454 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24848715
CAS Number: 1449008-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR