(6-Bromo-2-fluoro-3-iodophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

25945-1G 25945-5G 25945-25G
77341-564EA 93.73 USD
77341-564 77361-058 77372-830
(6-Bromo-2-fluoro-3-iodophenyl)methanol ≥97%
(6-Bromo-2-fluoro-3-iodophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₅BrFIO
Storage Temperature: Ambient
MDL Number: MFCD24848702
CAS Number: 1449008-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR