3-(Methoxymethoxy)-4-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

25870-1G 25870-5G 25870-25G
77341-186EA 74.16 USD
77341-186 77361-052 77372-824
3-(Methoxymethoxy)-4-methylbenzoic acid ≥95%
3-(Methoxymethoxy)-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
Storage Temperature: Ambient
MDL Number: MFCD21101772
CAS Number: 913392-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR