[1,1'-Biphenyl]-3-carboxaldehyde, 4'-fluoro-2',4-dimethoxy- ≥95%

Supplier: AOBChem USA

Warning

25866-500MG 25866-1G 25866-5G
77324-730EA 334.75 USD
77324-730 77348-584 77372-822
[1,1'-Biphenyl]-3-carboxaldehyde, 4'-fluoro-2',4-dimethoxy- ≥95%
[1,1'-Biphenyl]-3-carboxaldehyde, 4'-fluoro-2',4-dimethoxy-

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₃
MW: 260.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23713166
CAS Number: 408372-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR