4-Fluoro-7-(methylthio)-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

25864-500MG 25864-1G 25864-5G
77337-698EA 437.75 USD
77337-698 77348-582 77372-820
4-Fluoro-7-(methylthio)-1H-indazole ≥97%
4-Fluoro-7-(methylthio)-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇FN₂S
MW: 182.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23713165
CAS Number: 1428234-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR