4-(Methoxymethoxy)-3-(1-methylethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

25824-500MG 25824-1G 25824-5G
77324-724EA 341.96 USD
77324-724 77348-576 77372-816
4-(Methoxymethoxy)-3-(1-methylethyl)phenylboronic acid ≥97%
4-(Methoxymethoxy)-3-(1-methylethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₇BO₄
MW: 224.061 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25964665
CAS Number: 565452-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR