3-(5-Chloropyridin-2-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

25778-500MG 25778-1G 25778-5G 25778-25G
77324-720EA 199.82 USD
77324-720 77348-572 77361-040 77384-374
3-(5-Chloropyridin-2-yl)benzaldehyde ≥97%
3-(5-Chloropyridin-2-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₈ClNO
MW: 217.651 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17677898
CAS Number: 1255634-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR