3-Chloro-biphenyl-4-ylamine ≥97%

Supplier: AOBChem USA

Warning

25777-500MG 25777-1G 25777-5G 25777-25G
77324-718EA 88.58 USD
77324-718 77342-322 77361-038 77372-812
3-Chloro-biphenyl-4-ylamine ≥97%
3-Chloro-biphenyl-4-ylamine

Precaución: for Research Use Only

Formula: C₁₂H₁₀ClN
MW: 203.667 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01740412
CAS Number: 7285-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR