2,6-Difluoro-3-methylsulfanyl-benzoic acid methyl ester ≥95%

Supplier: AOBChem USA

Warning

25776-500MG 25776-1G 25776-5G 25776-25G
77324-716EA 218.36 USD
77324-716 77348-570 77361-036 77384-372
2,6-Difluoro-3-methylsulfanyl-benzoic acid methyl ester ≥95%
2,6-Difluoro-3-methylsulfanyl-benzoic acid methyl ester

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂S
MW: 218.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28977195
CAS Number: 2027537-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR