4-(2-Bromo-5-hydroxybenzyl)morpholine ≥95%

Supplier: AOBChem USA

Warning

25775-500MG 25775-1G 25775-5G 25775-25G
77324-714EA 201.88 USD
77324-714 77348-568 77361-034 77384-370
4-(2-Bromo-5-hydroxybenzyl)morpholine ≥95%
4-(2-Bromo-5-hydroxybenzyl)morpholine

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrNO₂
MW: 272.138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09836706
CAS Number: 937636-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR