2-Fluoro-3-methyl-5-(pyridin-4-yl)pyridine ≥97%

Supplier: AOBChem USA

Warning

25760-500MG 25760-1G 25760-25G 25760-5G
77324-706EA 289.43 USD
77324-706 77348-562 77384-364 77372-808
2-Fluoro-3-methyl-5-(pyridin-4-yl)pyridine ≥97%
2-Fluoro-3-methyl-5-(pyridin-4-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₉FN₂
MW: 188.201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15475652
CAS Number: 1214389-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR