2-Fluoro-3-methyl-5-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

25758-500MG 25758-1G 25758-25G 25758-5G
77324-702EA 289.43 USD
77324-702 77348-558 77384-360 77372-804
2-Fluoro-3-methyl-5-phenylpyridine ≥97%
2-Fluoro-3-methyl-5-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀FN
Storage Temperature: Ambient
MDL Number: MFCD14701872
CAS Number: 1214362-13-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR