5-(4-Chlorophenyl)-2-methylaniline ≥95%

Supplier: AOBChem USA

Warning

25719-500MG 25719-1G 25719-5G 25719-25G
77324-700EA 222.48 USD
77324-700 77348-556 77361-026 77384-358
5-(4-Chlorophenyl)-2-methylaniline ≥95%
5-(4-Chlorophenyl)-2-methylaniline

Precaución: for Research Use Only

Formula: C₁₃H₁₂ClN
Storage Temperature: Ambient
MDL Number: MFCD12779838
CAS Number: 244159-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR