N-(2,6-Dichloro-4-methylphenyl)acetamide ≥95%

Supplier: AOBChem USA

Warning

25707-500MG 25707-1G 25707-5G 25707-25G
77324-698EA 99.91 USD
77324-698 77342-756 77361-024 77372-802
N-(2,6-Dichloro-4-methylphenyl)acetamide ≥95%
N-(2,6-Dichloro-4-methylphenyl)acetamide

Precaución: for Research Use Only

Formula: C₉H₉Cl₂NO
MW: 218.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24549854
CAS Number: 99520-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR