4'-Fluoro-2'-methoxybiphenyl-3-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

25640-500MG 25640-1G 25640-5G
77335-628EA 361.53 USD
77335-628 77348-552 77372-798
4'-Fluoro-2'-methoxybiphenyl-3-carbaldehyde ≥95%
4'-Fluoro-2'-methoxybiphenyl-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁FO₂
MW: 230.234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23713138
CAS Number: 1329115-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR