2-Cyclopropyl-4-fluoroaniline ≥97%

Supplier: AOBChem USA

Warning

25621-500MG 25621-1G 25621-5G 25621-25G
77324-690EA 191.58 USD
77324-690 77348-544 77361-016 77384-354
2-Cyclopropyl-4-fluoroaniline ≥97%
2-Cyclopropyl-4-fluoroaniline

Precaución: for Research Use Only

Formula: C₉H₁₀FN
Storage Temperature: Ambient
MDL Number: MFCD16778828
CAS Number: 1250966-57-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR