5-Methyl-2-nitro-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

25620-500MG 25620-1G 25620-5G
77324-688EA 332.69 USD
77324-688 77348-542 77372-788
5-Methyl-2-nitro-1,1'-biphenyl ≥95%
5-Methyl-2-nitro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO₂
MW: 213.232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902568
CAS Number: 29547-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR