5-Methyl-2-phenylpyridine-3-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

25611-500MG 25611-1G 25611-5G 25611-25G
77324-686EA 199.82 USD
77324-686 77348-538 77361-014 77384-352
5-Methyl-2-phenylpyridine-3-carboxaldehyde ≥97%
5-Methyl-2-phenylpyridine-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO
MW: 197.233 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23713132
CAS Number: 1428234-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR