5-Chloro-2-phenylpyridine-4-methanol ≥95%

Supplier: AOBChem USA

Warning

25586-500MG 25586-1G 25586-5G
77337-474EA 415.09 USD
77337-474 77348-536 77372-784
5-Chloro-2-phenylpyridine-4-methanol ≥95%
5-Chloro-2-phenylpyridine-4-methanol

Precaución: for Research Use Only

Formula: C₁₂H₁₀ClNO
MW: 219.667 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16607284
CAS Number: 1227572-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR