3-(Methylthio)-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

25557-500MG 25557-1G 25557-5G
77335-530EA 357.41 USD
77335-530 77348-530 77372-774
3-(Methylthio)-5-(trifluoromethyl)benzaldehyde ≥97%
3-(Methylthio)-5-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₇F₃OS
MW: 220.211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23713129
CAS Number: 868166-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR