4-Ethoxy-2,3-dimethylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

25544-500MG 25544-1G 25544-5G 25544-25G
77324-682EA 263.68 USD
77324-682 77348-526 77361-006 77384-350
4-Ethoxy-2,3-dimethylbenzoic acid ≥95%
4-Ethoxy-2,3-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14537915
CAS Number: 1216009-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR