2-Fluoro-4-(methylthio)nicotinaldehyde ≥95%

Supplier: AOBChem USA

Warning

25522-500MG 25522-1G 25522-5G
77336-172EA 380.07 USD
77336-172 77348-524 77372-768
2-Fluoro-4-(methylthio)nicotinaldehyde ≥95%
2-Fluoro-4-(methylthio)nicotinaldehyde

Precaución: for Research Use Only

Formula: C₇H₆FNOS
MW: 171.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27756654
CAS Number: 1289172-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR