2'-Methoxy-1,1'-biphenyl-2-carbonitrile ≥97%

Supplier: AOBChem USA

Warning

25517-500MG 25517-1G 25517-5G
77337-536EA 418.18 USD
77337-536 77348-522 77372-766
2'-Methoxy-1,1'-biphenyl-2-carbonitrile ≥97%
2'-Methoxy-1,1'-biphenyl-2-carbonitrile

Precaución: for Research Use Only

Formula: C₁₄H₁₁NO
MW: 209.243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12403296
CAS Number: 150766-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR