2,6-Difluoro-4-(methylthio)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(2,6-Difluoro-4-(methylthio)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

25499-500MG 25499-1G 25499-5G
77324-680EA 317.24 USD
77324-680 77348-520 77372-764
2,6-Difluoro-4-(methylthio)phenylboronic acid pinacol ester ≥97%
2,6-Difluoro-4-(methylthio)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BF₂O₂S
MW: 286.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22493657
CAS Number: 1355011-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR