2,6-Difluoro-3-(methylthio)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

25496-500MG 25496-1G 25496-5G 25496-25G
77324-678EA 201.88 USD
77324-678 77348-518 77361-004 77384-348
2,6-Difluoro-3-(methylthio)benzaldehyde ≥95%
2,6-Difluoro-3-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆F₂OS
MW: 188.194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23713120
CAS Number: 1428234-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR