6-Bromo-2-chloro-3-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

25493-500MG 25493-1G 25493-5G 25493-25G
77324-676EA 154.5 USD
77324-676 77345-542 77361-002 77384-346
6-Bromo-2-chloro-3-methylbenzyl alcohol ≥97%
6-Bromo-2-chloro-3-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₈BrClO
Storage Temperature: Ambient
MDL Number: MFCD23713118
CAS Number: 1428234-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR