5-Chloro-3-(4-Chlorophenyl)-2-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

25460-500MG 25460-1G 25460-5G 25460-25G
77324-674EA 266.77 USD
77324-674 77348-516 77361-000 77384-344
5-Chloro-3-(4-Chlorophenyl)-2-fluoropyridine ≥97%
5-Chloro-3-(4-Chlorophenyl)-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₁₁H₆Cl₂FN
Storage Temperature: Ambient
MDL Number: MFCD23162310
CAS Number: 1364677-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR