3-Chloro-5-cylclopropyl-2-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

25441-500MG 25441-1G 25441-5G
77337-394EA 414.06 USD
77337-394 77348-514 77372-762
3-Chloro-5-cylclopropyl-2-methoxypyridine ≥97%
3-Chloro-5-cylclopropyl-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO
MW: 183.635 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564017
CAS Number: 2245949-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR