2-Ethoxy-5-(4-(trifluoromethoxy)phenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

25433-500MG 25433-1G 25433-5G
77335-626EA 361.53 USD
77335-626 77348-508 77372-758
2-Ethoxy-5-(4-(trifluoromethoxy)phenyl)pyridine ≥97%
2-Ethoxy-5-(4-(trifluoromethoxy)phenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₃NO₂
MW: 283.246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18404910
CAS Number: 121219-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR