3-Bromo-2-methoxy-5-phenylpyridine ≥95%

Supplier: AOBChem USA

Warning

25426-500MG 25426-1G 25426-5G
77335-528EA 357.41 USD
77335-528 77348-504 77372-754
3-Bromo-2-methoxy-5-phenylpyridine ≥95%
3-Bromo-2-methoxy-5-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO
Storage Temperature: Ambient
MDL Number: MFCD23713112
CAS Number: 1428234-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR