(6-Bromo-2,3,4-trifluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

25411-500MG 25411-1G 25411-5G
77336-054EA 375.95 USD
77336-054 77348-502 77372-750
(6-Bromo-2,3,4-trifluorophenyl)methanol ≥97%
(6-Bromo-2,3,4-trifluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₄BrF₃O
MW: 241.005 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27926975
CAS Number: 651326-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR