5-Chloro-2-fluoro-3-(4,4,5,5-tetramethyl-1,3-dioxolan-2-yl)pyridine ≥97%

Supplier: AOBChem USA

25345-500MG 25345-1G 25345-5G 25345-25G
77324-666EA 83.43 USD
77324-666 77342-186 77360-992 77372-748
5-Chloro-2-fluoro-3-(4,4,5,5-tetramethyl-1,3-dioxolan-2-yl)pyridine ≥97%
5-Chloro-2-fluoro-3-(4,4,5,5-tetramethyl-1,3-dioxolan-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₅ClFNO₂
MW: 259.704 g/mol
MDL Number: MFCD23713109
CAS Number: 1383446-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR