1-Methoxy-2-methyl-3,5-bis(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

25260-500MG 25260-1G 25260-5G
77324-658EA 346.08 USD
77324-658 77348-494 77372-744
1-Methoxy-2-methyl-3,5-bis(trifluoromethyl)benzene ≥95%
1-Methoxy-2-methyl-3,5-bis(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₈F₆O
MW: 258.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22628262
CAS Number: 180134-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR