1-Bromo-4-ethoxy-2,3-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

25227-500MG 25227-1G 25227-5G 25227-25G
77324-652EA 160.68 USD
77324-652 77345-758 77360-980 77384-328
1-Bromo-4-ethoxy-2,3-dimethylbenzene ≥97%
1-Bromo-4-ethoxy-2,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD23713101
CAS Number: 1428234-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR