(4-Bromo-2,3-dimethylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

25224-500MG 25224-1G 25224-5G 25224-25G
77324-650EA 222.48 USD
77324-650 77348-488 77360-978 77384-326
(4-Bromo-2,3-dimethylphenyl)methanol ≥95%
(4-Bromo-2,3-dimethylphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
MW: 215.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28977181
CAS Number: 1785359-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR