7-Chloro-6-fluoro-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

25219-500MG 25219-1G 25219-5G
77335-624EA 361.53 USD
77335-624 77348-486 77372-740
7-Chloro-6-fluoro-1H-indazole ≥95%
7-Chloro-6-fluoro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄ClFN₂
Storage Temperature: Ambient
MDL Number: MFCD22628260
CAS Number: 1414870-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR