2-Chloro-4'-methyl-biphenyl-4-ylamine ≥97%
Supplier: AOBChem USA
|
Warning
|
25216-500MG
25216-1G
25216-5G
25216-25G
77324-646EA
224.54
USD
77324-646
77348-484
77360-974
77384-322
2-Chloro-4'-methyl-biphenyl-4-ylamine ≥97%
2-Chloro-4'-methyl-biphenyl-4-ylamine
Precaución: for Research Use Only
Formula:
C₁₃H₁₂ClN MW: 217.694 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14585517 CAS Number: 1208087-74-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...