2-Chloro-4-(4-methylphenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

25210-500MG 25210-1G 25210-5G 25210-25G
77324-642EA 226.6 USD
77324-642 77348-482 77360-970 77384-318
2-Chloro-4-(4-methylphenyl)pyridine ≥97%
2-Chloro-4-(4-methylphenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀ClN
MW: 203.667 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22628258
CAS Number: 1202574-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR