2-Chloro-4-(4-trifluoromethylphenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

25209-500MG 25209-1G 25209-5G 25209-25G
77324-640EA 181.28 USD
77324-640 77346-780 77360-968 77384-316
2-Chloro-4-(4-trifluoromethylphenyl)pyridine ≥97%
2-Chloro-4-(4-trifluoromethylphenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₇ClF₃N
Storage Temperature: Ambient
MDL Number: MFCD08235167
CAS Number: 545396-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR