2-Methoxy-5-[4-(trifluoromethoxy)phenyl]pyridiine ≥97%

Supplier: AOBChem USA

Warning

25179-500MG 25179-1G 25179-5G
77324-636EA 334.75 USD
77324-636 77348-480 77372-738
2-Methoxy-5-[4-(trifluoromethoxy)phenyl]pyridiine ≥97%
2-Methoxy-5-[4-(trifluoromethoxy)phenyl]pyridiine

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₃NO₂
MW: 269.219 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18407628
CAS Number: 1261831-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR