4-Bromo-2,6-diflluoro-N,N-dimethyl-benzenemethanamine ≥97%

Supplier: AOBChem USA

Warning

25120-500MG 25120-1G 25120-5G 25120-25G
77324-632EA 121.54 USD
77324-632 77343-906 77360-960 77372-730
4-Bromo-2,6-diflluoro-N,N-dimethyl-benzenemethanamine ≥97%
4-Bromo-2,6-diflluoro-N,N-dimethyl-benzenemethanamine

Precaución: for Research Use Only

Formula: C₉H₁₀BrF₂N
Storage Temperature: Ambient
MDL Number: MFCD22628256
CAS Number: 1247006-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR