2-Chloro-2'-fluoro-3,4-bipyridine ≥97%

Supplier: AOBChem USA

Warning

25107-500MG 25107-1G 25107-5G
77336-170EA 380.07 USD
77336-170 77348-476 77372-728
2-Chloro-2'-fluoro-3,4-bipyridine ≥97%
2-Chloro-2'-fluoro-3,4-bipyridine

Precaución: for Research Use Only

Formula: C₁₀H₆ClFN₂
MW: 208.619 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13191924
CAS Number: 870221-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR