2-Chloro-N,N-diethylnicotinamide ≥97%

Supplier: AOBChem USA

Warning

25089-1G 25089-5G 25089-25G
77341-910EA 108.15 USD
77341-910 77360-958 77372-726
2-Chloro-N,N-diethylnicotinamide ≥97%
2-Chloro-N,N-diethylnicotinamide

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD03383605
CAS Number: 38029-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR