5-Cyclopropyl-3-ethoxypyridine ≥97%

Supplier: AOBChem USA

Warning

25083-500MG 25083-1G 25083-25G 25083-5G
77324-630EA 289.43 USD
77324-630 77348-474 77384-312 77372-724
5-Cyclopropyl-3-ethoxypyridine ≥97%
5-Cyclopropyl-3-ethoxypyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD22628254
CAS Number: 1414870-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR