2-Ethoxy-5-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

25081-500MG 25081-1G 25081-25G 25081-5G
77324-628EA 305.91 USD
77324-628 77348-472 77384-310 77372-722
2-Ethoxy-5-phenylpyridine ≥97%
2-Ethoxy-5-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO
MW: 199.248 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14701795
CAS Number: 136364-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR