2-Methoxy-5-phenylpyridine-4-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

25078-500MG 25078-1G 25078-5G 25078-25G
77324-626EA 222.48 USD
77324-626 77348-470 77360-956 77384-308
2-Methoxy-5-phenylpyridine-4-carboxaldehyde ≥97%
2-Methoxy-5-phenylpyridine-4-carboxaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD16606792
CAS Number: 1227582-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR