2-Fluoro-3-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

25075-500MG 25075-1G 25075-5G 25075-25G
77324-624EA 222.48 USD
77324-624 77348-468 77360-954 77384-306
2-Fluoro-3-phenylpyridine ≥97%
2-Fluoro-3-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₁H₈FN
Storage Temperature: Ambient
MDL Number: MFCD04114106
CAS Number: 361147-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR