Methyl 2,6-dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate ≥97%

Supplier: AOBChem USA

Warning

25068-500MG 25068-1G 25068-5G 25068-25G
77324-622EA 279.13 USD
77324-622 77348-466 77360-952 77384-304
Methyl 2,6-dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate ≥97%
Methyl 2,6-dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₄H₁₇BCl₂O₄
MW: 331 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20441900
CAS Number: 1321613-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR