4-Cyclopropyl-3,5-difluoroaniline ≥97%

Supplier: AOBChem USA

Warning

24972-500MG 24972-1G 24972-5G 24972-25G
77324-620EA 116.39 USD
77324-620 77343-480 77360-950 77372-720
4-Cyclopropyl-3,5-difluoroaniline ≥97%
4-Cyclopropyl-3,5-difluoroaniline

Precaución: for Research Use Only

Formula: C₉H₉F₂N
Storage Temperature: Ambient
MDL Number: MFCD14585542
CAS Number: 1208087-40-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR